siRNA Details
| Gene Symbol: SCRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145023.1 | NP_001138495.1 | 6 | 950 | ||
| NM_138355.3 | NP_612364.2 | 6 | 950 | |||
| XM_005257776.4 | XP_005257833.1 | 6 | 930 | |||
| XM_017025286.1 | XP_016880775.1 | 6 | 878 | |||
| GenBank mRNA | AK290646.1 | - | 6 | 853 | ||
| AK303835.1 | - | 6 | 906 | |||
| BC002980.1 | - | 6 | 893 | |||
| BC010408.2 | - | 6 | 864 | 3945715 | ||
| BC017317.2 | - | 6 | 863 | 3832079 | ||

SCRN2
Genomic Map