siRNA Details
| Gene Symbol: CEP95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316990.1 | NP_001303919.1 | 3, 4 | 626 | ||
| NM_138363.2 | NP_612372.1 | 3, 4 | 626 | |||
| NR_133644.1 | 4 | 645 | ||||
| XM_005257779.3 | XP_005257836.1 | 3, 4 | 602 | |||
| XM_005257780.2 | XP_005257837.1 | 3, 4 | 351 | |||
| XM_006722166.3 | XP_006722229.1 | 3, 4 | 368 | |||
| XM_011525458.2 | XP_011523760.1 | 3, 4 | 602 | |||
| XM_017025287.1 | XP_016880776.1 | 3, 4 | 370 | |||
| XM_017025288.1 | XP_016880777.1 | 3, 4 | 368 | |||
| GenBank mRNA | AK057326.1 | - | 3, 4 | 600 | ||
| AK290587.1 | - | 3, 4 | 366 | |||
| AK297409.1 | - | 3, 4 | 327 | |||
| AK316257.1 | - | 3, 4 | 370 | |||
| AL832822.1 | - | 4 | 296 | |||
| BC009518.1 | - | 3, 4 | 322 | 3633884 | ||

CEP95
Genomic Map