siRNA Details
| Gene Symbol: MTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138384.2 | NP_612393.2 | 11 | 1407 | ||
| GenBank mRNA | AK074976.1 | - | 10 | 1886 | ||
| AK095872.1 | - | 5 | 2276 | |||
| AK125012.1 | - | 11 | 2252 | |||
| BC000920.2 | - | 9 | 1116 | |||
| BC004409.1 | - | 11 | 1366 | |||
| BC026039.1 | - | 11 | 1379 | 5444745 | ||
| BC026725.1 | - | 11 | 1286 | |||
| BC035721.1 | - | 11 | 1311 | 5588933 | ||

MTG1
Genomic Map