siRNA Details
Gene Symbol: SGF29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138414.2 | NP_612423.1 | 10 | 1057 | ||
GenBank mRNA | AK057008.1 | - | 10 | 1016 | ||
BC011981.1 | - | 10 | 1048 | 3863665 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SGF29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138414.2 | NP_612423.1 | 10 | 1057 | ||
GenBank mRNA | AK057008.1 | - | 10 | 1016 | ||
BC011981.1 | - | 10 | 1048 | 3863665 |