siRNA Details
| Gene Symbol: LOC93622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026375.1 | - | 1 | 754 | ||
| BC021542.1 | - | 1 | 726 | |||
| BC066911.1 | - | 1 | 834 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC93622| Gene Symbol: LOC93622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026375.1 | - | 1 | 754 | ||
| BC021542.1 | - | 1 | 726 | |||
| BC066911.1 | - | 1 | 834 | |||