siRNA Details
| Gene Symbol: CALML6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138705.2 | NP_619650.2 | 3 | 579 | ||
| XM_005244729.3 | XP_005244786.2 | 3 | 1721 | |||
| XM_011540841.2 | XP_011539143.1 | 2 | 1604 | |||
| GenBank mRNA | AF490905.2 | - | 3 | 579 | ||
| AY566229.1 | - | 3 | 579 | |||

CALML6
Genomic Map