siRNA Details
Gene Symbol: GINM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138785.3 | NP_620140.1 | 1, 2 | 151 | ||
GenBank mRNA | AK289402.1 | - | 1, 2 | 138 | ||
AK295690.1 | - | 1, 2 | 177 | |||
AK308844.1 | - | 1, 2 | 138 | |||
AK315729.1 | - | 1, 2 | 148 | |||
AY358952.1 | - | 1, 2 | 269 | |||
BC014320.1 | - | 1, 2 | 155 | 3686673 |
