siRNA Details
| Gene Symbol: PLD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308174.1 | NP_001295103.1 | 8 | 1217 | ||
| NM_138790.3 | NP_620145.2 | 8 | 1179 | |||
| XM_011536411.2 | XP_011534713.1 | 8 | 1227 | |||
| XM_017020965.1 | XP_016876454.1 | 8 | 1116 | |||
| GenBank mRNA | AK295352.1 | - | 8 | 1172 | ||
| AK296188.1 | - | 8 | 1217 | |||
| AY358843.1 | - | 8 | 1145 | |||
| BC015003.1 | - | 8 | 1108 | 4853026 | ||

PLD4
Genomic Map