siRNA Details
| Gene Symbol: SRPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003137.4 | NP_003128.3 | 2, 3 | 203 | ||
| GenBank mRNA | AJ318054.1 | - | 1, 2 | 601 | ||
| AK292277.1 | - | 2, 3 | 148 | |||
| AK312712.1 | - | 2, 3 | 95 | |||
| BC038292.1 | - | 2, 3 | 168 | 4824261 | ||
| BG772464.1 | - | 2, 3 | 211 | |||
| BX648663.1 | - | 2, 3 | 120 | |||
| BX648697.1 | - | 2, 3 | 121 | |||
| DB233331.1 | - | 2, 3 | 203 | |||

SRPK1
Genomic Map