siRNA Details
| Gene Symbol: SMIM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135674.1 | NP_001129146.1 | 3 | 944 | ||
| NM_001135675.1 | NP_001129147.1 | 3 | 552 | |||
| NM_001135676.1 | NP_001129148.1 | 3 | 237 | |||
| NM_138436.3 | NP_612445.2 | 3 | 323 | |||
| XM_005273398.4 | XP_005273455.1 | 3 | 858 | |||
| GenBank mRNA | AK311932.1 | - | 3 | 298 | ||
| BC013035.2 | - | 3 | 285 | 3535608 | ||
| BI463786.1 | - | 3 | 242 | |||
| BI549696.1 | - | 3 | 557 | |||
| DA702519.1 | - | 3 | 323 | |||

SMIM19
Genomic Map