siRNA Details
| Gene Symbol: ZNF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308123.1 | NP_001295052.1 | 2 | 573 | ||
| NM_006965.3 | NP_008896.2 | 2 | 583 | |||
| XM_005258341.3 | XP_005258398.1 | 2 | 570 | |||
| GenBank mRNA | AF038964.1 | - | 2 | 527 | ||
| AF542097.1 | - | 1 | 404 | |||
| AK291246.1 | - | 2 | 562 | |||
| BC003566.1 | - | 2 | 533 | 3537160 | ||
| BC016801.1 | - | 2 | 554 | 4072599 | ||
| BT006658.1 | - | 1 | 392 | |||
| CR450328.1 | - | 1 | 392 | |||
| U68536.1 | - | 2 | 556 | |||

ZNF24
Genomic Map