siRNA Details
| Gene Symbol: TMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138691.2 | NP_619636.2 | 7 | 629 | ||
| XM_017014256.1 | XP_016869745.1 | 4 | 352 | |||
| GenBank mRNA | AF417578.1 | - | 7 | 629 | ||
| AK307461.1 | - | 6 | 479 | |||
| BC103955.1 | - | 3 | 117 | |||
| BC103956.1 | - | 3 | 117 | |||
| BC114380.1 | - | 3 | 117 | |||
| BC141860.1 | - | 3 | 117 | |||

TMC1
Genomic Map