siRNA Details
| Gene Symbol: KLF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138693.3 | NP_619638.2 | 1 | 1224 | ||
| GenBank mRNA | AF490374.1 | - | 1 | 1224 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLF14| Gene Symbol: KLF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138693.3 | NP_619638.2 | 1 | 1224 | ||
| GenBank mRNA | AF490374.1 | - | 1 | 1224 | ||