siRNA Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138771.3 | NP_620126.2 | 3, 4 | 697 | ||
| XM_017012775.1 | XP_016868264.1 | 4, 5 | 937 | |||
| XM_017012776.1 | XP_016868265.1 | 3, 4 | 680 | |||
| GenBank mRNA | AK026684.1 | - | 1, 2 | 194 | ||
| AK289911.1 | - | 3, 4 | 673 | |||
| AY358713.1 | - | 3, 4 | 664 | |||
| BC012427.1 | - | 1, 2 | 278 | 3858521 | ||
| BC104638.1 | - | 3, 4 | 589 | 6620343 | ||

CCDC126
Genomic Map