siRNA Details
| Gene Symbol: TM4SF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184723.1 | NP_001171652.1 | 1, 2 | 267 | ||
| NM_138786.3 | NP_620141.1 | 2, 3 | 443 | |||
| GenBank mRNA | AK313400.1 | - | 2, 3 | 217 | ||
| AL833134.1 | - | 2, 3 | 366 | |||
| BC014339.1 | - | 2, 3 | 319 | 3828466 | ||
| BF213446.1 | - | 278 | ||||
| CA454331.1 | - | 1, 2 | 229 | |||

TM4SF18
Genomic Map