siRNA Details
| Gene Symbol: ADPRHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138430.4 | NP_612439.2 | 5 | 794 | ||
| NM_199162.2 | NP_954631.1 | 5 | 637 | |||
| GenBank mRNA | AJ313429.1 | - | 5 | 794 | ||
| BC009755.2 | - | 5 | 637 | 3838063 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADPRHL1| Gene Symbol: ADPRHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138430.4 | NP_612439.2 | 5 | 794 | ||
| NM_199162.2 | NP_954631.1 | 5 | 637 | |||
| GenBank mRNA | AJ313429.1 | - | 5 | 794 | ||
| BC009755.2 | - | 5 | 637 | 3838063 | ||