siRNA Details
| Gene Symbol: DHRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136050.2 | NP_001129522.1 | 6 | 891 | ||
| NM_138452.2 | NP_612461.1 | 6 | 883 | |||
| GenBank mRNA | AF418205.1 | - | 6 | 827 | ||
| AK058159.1 | - | 6 | 883 | |||
| AK126383.1 | - | 2 | 2146 | |||
| AL833917.1 | - | 6 | 786 | |||
| BC014057.2 | - | 6 | 797 | 3622913 | ||
| BC015943.1 | - | 6 | 694 | 3922816 | ||
| BX247980.1 | - | 6 | 807 | |||

DHRS1
Genomic Map