siRNA Details
| Gene Symbol: ZNF653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138783.3 | NP_620138.2 | 7 | 1628 | ||
| GenBank mRNA | AK131037.1 | - | 5 | 1321 | ||
| AK314986.1 | - | 6 | 1659 | |||
| AY072704.1 | - | 7 | 1497 | |||
| BC014187.2 | - | 7 | 1524 | 4634585 | ||
| BC016816.1 | - | 7 | 1548 | 4094670 | ||

ZNF653
Genomic Map