siRNA Details
| Gene Symbol: SPPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139015.4 | NP_620584.2 | 2, 3 | 594 | ||
| XM_011537925.2 | XP_011536227.1 | 2, 3 | 124 | |||
| GenBank mRNA | AB252457.1 | - | 2, 3 | 186 | ||
| AJ345030.1 | - | 2, 3 | 162 | |||
| AJ420898.1 | - | 2, 3 | 130 | |||
| AK074916.1 | - | 1, 2 | 77 | |||
| AK223590.1 | - | 2, 3 | 590 | |||
| AK291161.1 | - | 2, 3 | 416 | |||
| AY169313.1 | - | 2, 3 | 368 | |||
| BC073910.1 | - | 2, 3 | 375 | 4538522 | ||
| BC101625.1 | - | 2, 3 | 189 | 8069131 | ||
| BC101627.1 | - | 2, 3 | 189 | 8069133 | ||
| BU932077.1 | - | 2, 3 | 594 | |||

SPPL3
Genomic Map