siRNA Details
| Gene Symbol: TSTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139246.4 | NP_640339.4 | 10 | 1841 | ||
| XM_011518312.2 | XP_011516614.1 | 9 | 1304 | |||
| XM_017014342.1 | XP_016869831.1 | 9 | 1366 | |||
| GenBank mRNA | AK056453.1 | - | 8 | 968 | ||
| AK056752.1 | - | 3 | 402 | |||
| AK128615.1 | - | 11 | 1507 | |||
| AK291199.1 | - | 10 | 1462 | |||
| BC022958.1 | - | 1 | 489 | |||
| BC035604.1 | - | 10 | 1488 | 4385905 | ||
| BX538308.1 | - | 10 | 1482 | |||
| BX648493.1 | - | 10 | 1480 | |||
| KU178877.1 | - | 9 | 1272 | |||
| KU178878.1 | - | 9 | 1244 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_173033, BC057893, AK155456, AK028258.

TSTD2
Genomic Map