siRNA Details
| Gene Symbol: SLC17A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145288.1 | NP_001138760.1 | 2 | 444 | ||
| NM_139319.2 | NP_647480.1 | 2 | 444 | |||
| GenBank mRNA | AJ459241.1 | - | 2 | 307 | ||
| AK128319.1 | - | 2 | 444 | |||
| BC117229.1 | - | 2 | 155 | 40125780 | ||
| BC143396.1 | - | 2 | 155 | 9051903 | ||

SLC17A8
Genomic Map