siRNA Details
| Gene Symbol: DPPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138815.3 | NP_620170.3 | 4 | 537 | ||
| XM_011512443.1 | XP_011510745.1 | 4 | 378 | |||
| XM_011512444.2 | XP_011510746.1 | 4 | 520 | |||
| XM_011512445.2 | XP_011510747.1 | 4 | 403 | |||
| GenBank mRNA | AY283672.1 | - | 4 | 539 | ||
| BC018070.1 | - | 4 | 415 | 4817054 | ||

DPPA2
Genomic Map