siRNA Details
Gene Symbol: DEPDC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139160.2 | NP_631899.2 | 1 | 373 | ||
GenBank mRNA | AF447588.1 | - | 1 | 373 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DEPDC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139160.2 | NP_631899.2 | 1 | 373 | ||
GenBank mRNA | AF447588.1 | - | 1 | 373 |