siRNA Details
| Gene Symbol: ADCY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198568.1 | NP_001185497.1 | 4, 5 | 1096 | ||
| NM_001198592.1 | NP_001185521.1 | 5, 6 | 795 | |||
| NM_139247.3 | NP_640340.2 | 5, 6 | 791 | |||
| GenBank mRNA | AF497516.1 | - | 4, 5 | 683 | ||
| AK122714.1 | - | 5, 6 | 791 | |||
| AK126468.1 | - | 4, 5 | 645 | |||
| BC117473.1 | - | 4, 5 | 772 | 40126024 | ||
| BC117475.1 | - | 4, 5 | 772 | 40126026 | ||
| BC143947.1 | - | 4, 5 | 772 | |||
| BX248285.1 | - | 5, 6 | 762 | |||
| BX537489.1 | - | 1 | 1136 | |||

ADCY4
Genomic Map