siRNA Details
| Gene Symbol: TRIM43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138800.1 | NP_620155.1 | 7 | 1489 | ||
| GenBank mRNA | BC015353.1 | - | 7 | 1489 | 3445177 | |
| Gene Symbol: TRIM43B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164464.1 | NP_001157936.1 | 7 | 1483 | ||
| XM_011511669.1 | XP_011509971.1 | 7 | 1427 | |||

TRIM43
Genomic Map