siRNA Details
| Gene Symbol: DPPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138815.3 | NP_620170.3 | 7 | 1069 | ||
| XM_011512443.1 | XP_011510745.1 | 7 | 910 | |||
| XM_011512444.2 | XP_011510746.1 | 7 | 1052 | |||
| XM_011512445.2 | XP_011510747.1 | 7 | 935 | |||
| GenBank mRNA | AY283672.1 | - | 7 | 1071 | ||
| BC018070.1 | - | 7 | 947 | 4817054 | ||

DPPA2
Genomic Map