siRNA Details
| Gene Symbol: MASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031849.2 | NP_001027019.1 | 5 | 1085 | ||
| NM_001879.5 | NP_001870.3 | 5 | 1085 | |||
| NM_139125.3 | NP_624302.1 | 5 | 1085 | |||
| NR_033519.1 | 4 | 853 | ||||
| XM_006713701.2 | XP_006713764.1 | 6 | 1158 | |||
| XM_011512989.2 | XP_011511291.1 | 5 | 1079 | |||
| XM_011512990.2 | XP_011511292.1 | 5 | 1079 | |||
| XM_011512991.2 | XP_011511293.1 | 5 | 1074 | |||
| XM_017006869.1 | XP_016862358.1 | 6 | 1163 | |||
| XM_017006870.1 | XP_016862359.1 | 4 | 1051 | |||
| XM_017006871.1 | XP_016862360.1 | 5 | 1079 | |||
| XM_017006872.1 | XP_016862361.1 | 5 | 938 | |||
| GenBank mRNA | AF284421.1 | - | 5 | 785 | ||
| AK129893.1 | - | 5 | 784 | |||
| AK291157.1 | - | 5 | 921 | |||
| AK300663.1 | - | 6 | 906 | |||
| AK304334.1 | - | 4 | 563 | |||
| BC039724.1 | - | 5 | 1020 | |||
| BC106945.1 | - | 5 | 745 | 40034551 | ||
| BC106946.1 | - | 5 | 745 | 40034552 | ||
| BX647237.1 | - | 4 | 568 | |||
| CB162101.1 | - | 3 | 310 | |||
| CR749615.1 | - | 5 | 987 | |||

MASP1
Genomic Map