siRNA Details
| Gene Symbol: ANKRD22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144590.2 | NP_653191.2 | 2 | 527 | ||
| GenBank mRNA | AK313969.1 | - | 2 | 299 | ||
| BC021671.1 | - | 2 | 314 | 3682099 | ||
| BM785517.1 | - | 2 | 370 | |||
| DA672161.1 | - | 2 | 527 | |||

ANKRD22
Genomic Map