siRNA Details
| Gene Symbol: RFTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055136.1 | - | 3 | 775 | ||
| BC093966.1 | - | 3 | 422 | 7939811 | ||
| BC111069.1 | - | 3 | 907 | 5764096 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RFTN2| Gene Symbol: RFTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055136.1 | - | 3 | 775 | ||
| BC093966.1 | - | 3 | 422 | 7939811 | ||
| BC111069.1 | - | 3 | 907 | 5764096 | ||