siRNA Details
| Gene Symbol: FAM131A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171093.1 | NP_001164564.1 | 6 | 939 | ||
| NM_144635.4 | NP_653236.3 | 6 | 1057 | |||
| XM_005247113.2 | XP_005247170.1 | 7 | 1087 | |||
| XM_005247114.3 | XP_005247171.1 | 5 | 883 | |||
| XM_011512413.2 | XP_011510715.1 | 5 | 1125 | |||
| GenBank mRNA | AY358435.1 | - | 3 | 2185 | ||
| BC026221.1 | - | 5 | 696 | 4399325 | ||
| BC037299.1 | - | 6 | 936 | 4825140 | ||

FAM131A
Genomic Map