siRNA Details
| Gene Symbol: TMEM42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144638.2 | NP_653239.1 | 1, 2 | 216 | ||
| GenBank mRNA | BC019851.1 | - | 1, 2 | 209 | 5114515 | |
| DB499198.1 | - | 1, 2 | 216 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM42| Gene Symbol: TMEM42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144638.2 | NP_653239.1 | 1, 2 | 216 | ||
| GenBank mRNA | BC019851.1 | - | 1, 2 | 209 | 5114515 | |
| DB499198.1 | - | 1, 2 | 216 | |||