siRNA Details
| Gene Symbol: PPM1M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144641.3 | NP_653242.3 | 3 | 588 | ||
| XM_005264879.2 | XP_005264936.1 | 3 | 656 | |||
| XM_005264881.1 | XP_005264938.1 | 3 | 477 | |||
| GenBank mRNA | AB474372.1 | - | 3 | 556 | ||
| AK056894.1 | - | 3 | 477 | |||
| AK096681.1 | - | 3 | 483 | |||
| AL832484.1 | - | 2 | 399 | |||
| DA869388.1 | - | 3 | 478 | |||

PPM1M
Genomic Map