siRNA Details
| Gene Symbol: LINC00477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029451.2 | 1 | 504 | |||
| GenBank mRNA | AK057456.1 | - | 1 | 504 | ||
| BC104436.1 | - | 1 | 484 | |||
| BC104437.1 | - | 1 | 484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00477| Gene Symbol: LINC00477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029451.2 | 1 | 504 | |||
| GenBank mRNA | AK057456.1 | - | 1 | 504 | ||
| BC104436.1 | - | 1 | 484 | |||
| BC104437.1 | - | 1 | 484 | |||