siRNA Details
| Gene Symbol: OTOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161683.1 | NP_001155155.1 | 8 | 1146 | ||
| NM_144672.3 | NP_653273.3 | 12 | 1290 | |||
| NM_170664.2 | NP_733764.1 | 3 | 384 | |||
| XM_011545747.2 | XP_011544049.1 | 13 | 1669 | |||
| XM_011545748.2 | XP_011544050.2 | 2 | 663 | |||
| XM_017022950.1 | XP_016878439.1 | 13 | 1393 | |||
| GenBank mRNA | AK093062.1 | - | 3 | 384 | ||
| AK125840.1 | - | 8 | 1146 | |||
| AK302733.1 | - | 2 | 446 | |||
| BC129992.1 | - | 12 | 1287 | |||
| BC129993.1 | - | 12 | 1294 | 40128985 | ||

OTOA
Genomic Map