siRNA Details
| Gene Symbol: COQ10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099337.1 | NP_001092807.1 | 3 | 319 | ||
| NM_144576.3 | NP_653177.3 | 3 | 563 | |||
| GenBank mRNA | AK057003.1 | - | 3 | 479 | ||
| AK057014.1 | - | 2 | 280 | |||
| AY358861.1 | - | 3 | 251 | |||
| BC002435.2 | - | 2 | 140 | |||
| BC026922.1 | - | 3 | 319 | |||
| BC047444.1 | - | 3 | 291 | |||
| BC073923.1 | - | 3 | 447 | 5188138 | ||

COQ10A
Genomic Map