siRNA Details
| Gene Symbol: GNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297571.1 | NP_001284500.1 | 10 | 1264 | ||
| NM_002075.3 | NP_002066.1 | 10 | 1267 | |||
| XM_011520953.2 | XP_011519255.1 | 9 | 2188 | |||
| GenBank mRNA | AB209283.1 | - | 9 | 2408 | ||
| AB593098.1 | - | 10 | 1193 | |||
| AB593099.1 | - | 10 | 1192 | |||
| BC000115.1 | - | 10 | 1180 | 3351520 | ||
| BC002454.2 | - | 10 | 1187 | 3342882 | ||
| BC015920.1 | - | 10 | 1017 | 4648399 | ||
| M31328.1 | - | 9 | 1063 | |||

GNB3
Genomic Map