siRNA Details
| Gene Symbol: ZC3H18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294340.1 | NP_001281269.1 | 2 | 391 | ||
| NM_144604.3 | NP_653205.3 | 2 | 391 | |||
| XM_011522863.1 | XP_011521165.1 | 2 | 391 | |||
| XM_017022934.1 | XP_016878423.1 | 2 | 391 | |||
| GenBank mRNA | AK225807.1 | - | 2 | 361 | ||
| AK300254.1 | - | 2 | 369 | |||
| AK302716.1 | - | 2 | 369 | |||
| BC050463.1 | - | 2 | 338 | 6042036 | ||
| CN299704.1 | - | 1 | 25 | |||
| CN299707.1 | - | 2 | 350 | |||
| DA313229.1 | - | 2 | 391 | |||

ZC3H18
Genomic Map