siRNA Details
| Gene Symbol: C9orf116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048265.1 | NP_001041730.1 | 3 | 521 | ||
| NM_144654.2 | NP_653255.1 | 3 | 475 | |||
| GenBank mRNA | AK024007.1 | - | 3 | 1078 | ||
| BC021261.1 | - | 4 | 581 | 4136397 | ||
| DR003074.1 | - | 3 | 606 | |||

C9orf116
Genomic Map