siRNA Details
| Gene Symbol: ZC3H18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294340.1 | NP_001281269.1 | 19 | 3640 | ||
| NM_144604.3 | NP_653205.3 | 18 | 3568 | |||
| XM_011522863.1 | XP_011521165.1 | 17 | 3253 | |||
| XM_011522865.1 | XP_011521167.1 | 11 | 2010 | |||
| GenBank mRNA | AK026317.1 | - | 4 | 1095 | ||
| BC001584.1 | - | 15 | 2664 | |||
| BC050463.1 | - | 18 | 3515 | 6042036 | ||

ZC3H18
Genomic Map