siRNA Details
| Gene Symbol: LYZL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304386.1 | NP_001291315.1 | 5 | 860 | ||
| NM_144634.3 | NP_653235.1 | 5 | 879 | |||
| XM_017005705.1 | XP_016861194.1 | 7 | 1234 | |||
| XM_017005706.1 | XP_016861195.1 | 6 | 992 | |||
| GenBank mRNA | AF326749.1 | - | 5 | 808 | ||
| BC016747.2 | - | 5 | 850 | 4830396 | ||

LYZL4
Genomic Map