siRNA Details
| Gene Symbol: SCLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144643.3 | NP_653244.2 | 5, 6 | 802 | ||
| XM_017007716.1 | XP_016863205.1 | 5, 6 | 802 | |||
| XM_017007717.1 | XP_016863206.1 | 5, 6 | 802 | |||
| XM_017007718.1 | XP_016863207.1 | 5, 6 | 802 | |||
| XM_017007719.1 | XP_016863208.1 | 5, 6 | 323 | |||
| GenBank mRNA | AK055217.1 | - | 5, 6 | 802 | ||
| BC121057.1 | - | 5, 6 | 608 | |||
| BC128051.1 | - | 5, 6 | 607 | |||

SCLT1
Genomic Map