siRNA Details
| Gene Symbol: JCHAIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144646.3 | NP_653247.1 | 4 | 966 | ||
| XM_006714211.2 | XP_006714274.1 | 5 | 931 | |||
| XM_006714212.2 | XP_006714275.1 | 5 | 868 | |||
| XM_011531925.1 | XP_011530227.1 | 5 | 868 | |||
| XM_011531926.1 | XP_011530228.1 | 5 | 868 | |||
| GenBank mRNA | BC038982.1 | - | 4 | 883 | 5755337 | |
| BP221304.1 | - | 1 | 115 | |||
| BX648421.1 | - | 1 | 2426 | |||

JCHAIN
Genomic Map