siRNA Details
| Gene Symbol: ZNF354A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324339.1 | NP_001311268.1 | 4 | 463 | ||
| NM_005649.2 | NP_005640.2 | 5 | 547 | |||
| XM_011534645.2 | XP_011532947.1 | 5 | 1054 | |||
| XM_017009791.1 | XP_016865280.1 | 5 | 452 | |||
| GenBank mRNA | AF116030.1 | - | 5 | 553 | ||
| AK290711.1 | - | 5 | 442 | |||
| BC047105.1 | - | 5 | 504 | 5312219 | ||
| D89928.1 | - | 5 | 501 | |||
| EU794664.1 | - | 5 | 547 | |||

ZNF354A
Genomic Map