siRNA Details
| Gene Symbol: FAM109A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177996.1 | NP_001171467.1 | 4 | 2101 | ||
| NM_001177997.1 | NP_001171468.1 | 3 | 1994 | |||
| NM_144671.4 | NP_653272.2 | 3 | 2013 | |||
| GenBank mRNA | AK056918.1 | - | 3 | 1399 | ||
| AK093052.1 | - | 2 | 1717 | |||
| BC014091.2 | - | 4 | 728 | |||
| BC018996.2 | - | 4 | 728 | |||
| BC034809.1 | - | 3 | 1914 | 4943538 | ||

FAM109A
Genomic Map