siRNA Details
| Gene Symbol: ZNF480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297624.1 | NP_001284553.1 | 3 | 228 | ||
| NM_144684.3 | NP_653285.2 | 3 | 228 | |||
| XM_011526465.2 | XP_011524767.1 | 3 | 630 | |||
| GenBank mRNA | AK096559.1 | - | 3 | 218 | ||
| AK303271.1 | - | 3 | 219 | |||
| AY512662.1 | - | 3 | 197 | |||
| BC007056.1 | - | 3 | 223 | |||
| BC018783.1 | - | 3 | 197 | 4873910 | ||
| BC054018.1 | - | 3 | 228 | |||
| BC065503.1 | - | 3 | 203 | |||
| DA501214.1 | - | 3 | 226 | |||

ZNF480
Genomic Map