siRNA Details
| Gene Symbol: TMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145303.2 | NP_001138775.2 | 2 | 353 | ||
| NM_144686.3 | NP_653287.2 | 2 | 335 | |||
| XM_011526486.2 | XP_011524788.1 | 2 | 318 | |||
| GenBank mRNA | AK314973.1 | - | 2 | 256 | ||
| AY236492.1 | - | 2 | 352 | |||
| AY263166.1 | - | 2 | 232 | |||
| BC025323.1 | - | 2 | 242 | 2959382 | ||
| BQ082417.1 | - | 2 | 245 | |||
| DQ656026.2 | - | 2 | 181 | |||

TMC4
Genomic Map