siRNA Details
| Gene Symbol: PEBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144962.2 | NP_659399.2 | 2, 3 | 232 | ||
| XM_011544414.2 | XP_011542716.1 | 2, 3 | 287 | |||
| GenBank mRNA | AY037148.1 | - | 2, 3 | 205 | ||
| AY359109.1 | - | 2, 3 | 228 | |||
| AY730275.1 | - | 2, 3 | 183 | |||
| BC020779.1 | - | 2, 3 | 224 | 4700840 | ||
| BF853000.1 | - | 2, 3 | 232 | |||
| FJ224332.1 | - | 2, 3 | 224 | |||

PEBP4
Genomic Map