siRNA Details
| Gene Symbol: SLFN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144975.3 | NP_659412.3 | 2 | 302 | ||
| XM_005257934.3 | XP_005257991.1 | 2 | 343 | |||
| GenBank mRNA | AK054668.1 | - | 2 | 297 | ||
| AK303299.1 | - | 2 | 297 | |||
| AK303632.1 | - | 2 | 299 | |||
| BC021238.1 | - | 2 | 280 | |||
| BC125200.1 | - | 1 | 238 | 40123369 | ||
| BC125201.1 | - | 1 | 238 | 40123372 | ||
| BP281007.1 | - | 2 | 302 | |||
| BX647942.1 | - | 2 | 299 | |||

SLFN5
Genomic Map