siRNA Details
| Gene Symbol: ZFC3H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144982.4 | NP_659419.3 | 2 | 981 | ||
| GenBank mRNA | AB011118.2 | - | 2 | 831 | ||
| BC015679.2 | - | 2 | 870 | |||
| BC064336.1 | - | 2 | 862 | |||
| BC073843.1 | - | 2 | 836 | |||
| BX647709.1 | - | 2 | 861 | |||

ZFC3H1
Genomic Map