siRNA Details
| Gene Symbol: VWA3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144992.4 | NP_659429.4 | 14 | 2200 | ||
| XM_005263897.2 | XP_005263954.1 | 14 | 2165 | |||
| XM_006712357.2 | XP_006712420.1 | 13 | 1914 | |||
| XM_006712359.3 | XP_006712422.1 | 5 | 594 | |||
| XM_006712360.3 | XP_006712423.1 | 2 | 241 | |||
| XM_011510770.1 | XP_011509072.1 | 14 | 2165 | |||
| XM_011510771.2 | XP_011509073.1 | 13 | 2039 | |||
| XM_011510772.1 | XP_011509074.1 | 14 | 2165 | |||
| XM_011510774.1 | XP_011509076.1 | 14 | 2165 | |||
| XM_017003560.1 | XP_016859049.1 | 8 | 1031 | |||
| XM_017003561.1 | XP_016859050.1 | 8 | 1037 | |||
| XM_017003562.1 | XP_016859051.1 | 8 | 1087 | |||
| XM_017003563.1 | XP_016859052.1 | 8 | 1037 | |||
| XM_017003564.1 | XP_016859053.1 | 14 | 2165 | |||
| GenBank mRNA | AK057248.1 | - | 11 | 1701 | ||
| AK090786.1 | - | 2 | 167 | |||
| AK093084.1 | - | 2 | 167 | |||
| AK295743.1 | - | 8 | 1095 | |||
| AL832635.2 | - | 15 | 2143 | |||
| AL832761.1 | - | 14 | 2221 | |||
| AL834173.1 | - | 5 | 599 | |||
| AL834385.1 | - | 4 | 463 | |||
| BC026091.2 | - | 14 | 2015 | |||
| BC095480.1 | - | 15 | 2376 | |||
| BC150653.1 | - | 14 | 2140 | 9057024 | ||

VWA3B
Genomic Map